Assay Details
| Gene Symbol: HRASLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981407.2 | XP_008979655.1 | 2 - 3 | 837 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HRASLS| Gene Symbol: HRASLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981407.2 | XP_008979655.1 | 2 - 3 | 837 | 71 | |
Set Membership: |
|