Assay Details
| Gene Symbol: SLC30A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985266.2 | XP_008983514.2 | 1 - 2 | 786 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC30A1| Gene Symbol: SLC30A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985266.2 | XP_008983514.2 | 1 - 2 | 786 | 103 | |
Set Membership: |
|