Assay Details
| Gene Symbol: DNAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743783.4 | XP_002743829.1 | 2 - 3 | 338 | 76 | |
| XM_008979319.2 | XP_008977567.1 | 3 - 4 | 546 | 76 | ||
| XM_008979322.2 | XP_008977570.1 | 3 - 4 | 414 | 76 | ||
| XM_008979327.2 | XP_008977575.1 | 2 - 3 | 338 | 76 | ||
| XM_008979332.2 | XP_008977580.1 | 3 - 4 | 519 | 76 | ||
| XM_008979345.2 | XP_008977593.1 | 3 - 4 | 543 | 76 | ||
| XM_008979354.2 | XP_008977602.1 | 3 - 4 | 534 | 76 | ||
| XM_008979356.2 | XP_008977604.1 | 3 - 4 | 411 | 76 | ||
| XM_017963792.1 | XP_017819281.1 | 3 - 4 | 537 | 76 | ||
More Information
Set Membership: |
|

DNAL4
Genomic Map