Assay Details
| Gene Symbol: DNAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743783.4 | XP_002743829.1 | 3 - 4 | 422 | 92 | |
| XM_008979319.2 | XP_008977567.1 | 4 - 5 | 630 | 92 | ||
| XM_008979322.2 | XP_008977570.1 | 4 - 5 | 498 | 92 | ||
| XM_008979327.2 | XP_008977575.1 | 3 - 4 | 422 | 92 | ||
| XM_008979332.2 | XP_008977580.1 | 4 - 5 | 603 | 92 | ||
| XM_008979345.2 | XP_008977593.1 | 4 - 5 | 627 | 92 | ||
| XM_008979354.2 | XP_008977602.1 | 4 - 5 | 618 | 92 | ||
| XM_008979356.2 | XP_008977604.1 | 4 - 5 | 495 | 92 | ||
| XM_017963792.1 | XP_017819281.1 | 4 - 5 | 621 | 92 | ||
More Information
Set Membership: |
|

DNAL4
Genomic Map