Assay Details
| Gene Symbol: MED27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732067.3 | XP_003732115.2 | 2 - 3 | 386 | 86 | |
| XM_017976970.1 | XP_017832459.1 | 2 - 3 | 386 | 86 | ||
| XM_017976971.1 | XP_017832460.1 | 2 - 3 | 387 | 86 | ||
| XM_017976979.1 | XP_017832468.1 | 2 - 3 | 371 | 86 | ||
More Information
Set Membership: |
|

MED27
Genomic Map