Assay Details
Gene Symbol: TTC33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745027.3 | XP_002745073.1 | 2 - 3 | 362 | 72 | |
XM_008992131.2 | XP_008990379.1 | 2 - 3 | 348 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
