Assay Details
| Gene Symbol: SETBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963440.1 | XP_017818929.1 | 3 - 4 | 872 | 78 | |
| XM_017963441.1 | XP_017818930.1 | 3 - 4 | 1272 | 78 | ||
| XM_017963442.1 | XP_017818931.1 | 3 - 4 | 852 | 78 | ||
| XM_017963443.1 | XP_017818932.1 | 3 - 4 | 872 | 78 | ||
More Information
Set Membership: |
|

SETBP1
Genomic Map