Assay Details
| Gene Symbol: AMZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997636.2 | XP_008995884.1 | 7 - 8 | 1335 | 79 | |
| XM_008997637.2 | XP_008995885.1 | 8 - 9 | 1188 | 79 | ||
| XM_008997639.2 | XP_008995887.1 | 7 - 8 | 1136 | 79 | ||
| XM_008997640.2 | XP_008995888.1 | 7 - 8 | 1113 | 79 | ||
More Information
Set Membership: |
|

AMZ2
Genomic Map