Assay Details
| Gene Symbol: ARSF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762591.4 | XP_002762637.2 | 6 - 7 | 1442 | 71 | |
| XM_008988894.2 | XP_008987142.1 | 6 - 7 | 1600 | 71 | ||
| XM_008988896.2 | XP_008987144.1 | 5 - 6 | 855 | 71 | ||
| XM_008988898.2 | XP_008987146.1 | 5 - 6 | 1475 | 71 | ||
| XM_008988900.2 | XP_008987148.1 | 4 - 5 | 1324 | 71 | ||
More Information
Set Membership: |
|

ARSF
Genomic Map