Assay Details
Gene Symbol: STOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745183.4 | XP_002745229.1 | 2 - 3 | 691 | 83 | |
XM_008992402.2 | XP_008990650.1 | 2 - 3 | 693 | 83 | ||
XM_017970117.1 | XP_017825606.1 | 2 - 3 | 693 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
