Assay Details
| Gene Symbol: HAND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751878.3 | XP_002751924.1 | 1 | 190 | 86 | |
| XM_009002904.2 | XP_009001152.1 | 1 | 190 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002751878.3, XM_009002904.2 transcripts.
Set Membership: |
|

HAND1
Genomic Map