Assay Details
Gene Symbol: ANAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009006283.2 | XP_009004531.1 | 13 - 14 | 2274 | 62 | |
GenBank mRNA | EF215032.1 | - | 1 - 2 | 135 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
