Assay Details
| Gene Symbol: MAP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745394.3 | XP_002745440.1 | 13 - 14 | 2082 | 108 | |
| GenBank mRNA | EF215121.1 | - | 2 - 3 | 240 | 108 | |
More Information
Set Membership: |
|

MAP9
Genomic Map