Assay Details
| Gene Symbol: PCBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756263.3 | XP_002756309.1 | 3 - 4 | 464 | 86 | |
| XM_009009777.2 | XP_009008025.1 | 3 - 4 | 691 | 86 | ||
| XM_009009778.1 | XP_009008026.1 | 3 - 4 | 258 | 86 | ||
More Information
Set Membership: |
|

PCBD1
Genomic Map