Assay Details
| Gene Symbol: TIMMDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758780.4 | XP_002758826.2 | 6 - 7 | 939 | 69 | |
| XM_008982397.2 | XP_008980645.1 | 7 - 8 | 962 | 69 | ||
More Information
Set Membership: |
|

TIMMDC1
Genomic Map