Assay Details
| Gene Symbol: NDUFS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744985.4 | XP_002745031.1 | 1 - 2 | 137 | 68 | |
| XM_008992076.2 | XP_008990324.1 | 1 - 2 | 142 | 68 | ||
| XM_017969926.1 | XP_017825415.1 | 1 - 2 | 140 | 68 | ||
| XM_017969927.1 | XP_017825416.1 | 1 - 2 | 140 | 68 | ||
More Information
Set Membership: |
|

NDUFS4
Genomic Map