Assay Details
| Gene Symbol: NHLH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984734.2 | XP_008982982.1 | 2 | 1643 | 93 | |
| XM_017966401.1 | XP_017821890.1 | 3 | 1684 | 93 | ||
More Information
Set Membership: |
|

NHLH1
Genomic Map