Assay Details
| Gene Symbol: DUOXA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005367.2 | XP_009003615.1 | 1 | 410 | 82 | |
| XM_017976824.1 | XP_017832313.1 | 1 | 409 | 82 | ||
More Information
Set Membership: |
|

DUOXA2
Genomic Map