Assay Details
| Gene Symbol: PTGER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745028.3 | XP_002745074.1 | 3 | 3802 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTGER4| Gene Symbol: PTGER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745028.3 | XP_002745074.1 | 3 | 3802 | 100 | |
Set Membership: |
|