Assay Details
| Gene Symbol: PPIL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746990.4 | XP_002747036.1 | 5 - 6 | 687 | 86 | |
| XM_002746991.3 | XP_002747037.1 | 5 - 6 | 687 | 86 | ||
| XM_017971302.1 | XP_017826791.1 | 5 - 6 | 687 | 86 | ||
| XM_017971303.1 | XP_017826792.1 | 5 - 6 | 676 | 86 | ||
| XM_017971304.1 | XP_017826793.1 | 4 - 5 | 1016 | 86 | ||
| XM_017971305.1 | XP_017826794.1 | 5 - 6 | 687 | 86 | ||
| XM_017971306.1 | XP_017826795.1 | 5 - 6 | 687 | 86 | ||
| XM_017971307.1 | XP_017826796.1 | 5 - 6 | 687 | 86 | ||
More Information
Set Membership: |
|

PPIL6
Genomic Map