Assay Details
| Gene Symbol: TYRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017973584.1 | XP_017829073.1 | 5 - 6 | 1020 | 84 | |
| XM_017973585.1 | XP_017829074.1 | 6 - 7 | 977 | 84 | ||
More Information
Set Membership: |
|

TYRP1
Genomic Map