Assay Details
| Gene Symbol: LOC100411008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759123.4 | XP_002759169.1 | 3 - 4 | 364 | 75 | |
| XM_017965467.1 | XP_017820956.1 | 3 - 4 | 257 | 75 | ||
More Information
Set Membership: |
|

LOC100411008
Genomic Map