Assay Details
| Gene Symbol: CREB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751441.3 | XP_002751487.1 | 8 - 9 | 1412 | 56 | |
| XM_009002226.2 | XP_009000474.1 | 4 - 5 | 2609 | 56 | ||
| XM_017975229.1 | XP_017830718.1 | 9 - 10 | 1209 | 56 | ||
| XM_017975230.1 | XP_017830719.1 | 8 - 9 | 1088 | 56 | ||
| XM_017975231.1 | XP_017830720.1 | 8 - 9 | 3531 | 56 | ||
| XM_017975232.1 | XP_017830721.1 | 8 - 9 | 3531 | 56 | ||
More Information
Set Membership: |
|

CREB5
Genomic Map