Assay Details
| Gene Symbol: LOC100387657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009375.2 | XP_009007623.1 | 10 - 11 | 1159 | 62 | |
| XM_009009376.2 | XP_009007624.1 | 10 - 11 | 1134 | 62 | ||
| XM_009009377.2 | XP_009007625.1 | 10 - 11 | 1135 | 62 | ||
| XM_017978834.1 | XP_017834323.1 | 10 - 11 | 1131 | 62 | ||
More Information
Set Membership: |
|

LOC100387657
Genomic Map