Assay Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745106.3 | XP_002745152.1 | 6 - 7 | 1074 | 65 | |
| XM_017969984.1 | XP_017825473.1 | 7 - 8 | 1055 | 65 | ||
| XM_017969985.1 | XP_017825474.1 | 5 - 6 | 2957 | 65 | ||
More Information
Set Membership: |
|

OTULIN
Genomic Map