Assay Details
| Gene Symbol: GMNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746173.4 | XP_002746219.1 | 4 - 5 | 489 | 72 | |
| XM_008993884.2 | XP_008992132.1 | 4 - 5 | 596 | 72 | ||
| XM_008993885.2 | XP_008992133.1 | 4 - 5 | 540 | 72 | ||
| XM_008993886.2 | XP_008992134.1 | 4 - 5 | 570 | 72 | ||
More Information
Set Membership: |
|

GMNN
Genomic Map