Assay Details
| Gene Symbol: CDKN2AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745186.4 | XP_002745232.1 | 1 - 2 | 736 | 97 | |
| XM_008992405.2 | XP_008990653.1 | 1 - 2 | 397 | 97 | ||
| XM_017970121.1 | XP_017825610.1 | 1 - 2 | 736 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008992405.2 transcripts.
Set Membership: |
|

CDKN2AIP
Genomic Map