Assay Details
| Gene Symbol: NPTXR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963814.1 | XP_017819303.1 | 2 - 3 | 997 | 98 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPTXR| Gene Symbol: NPTXR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963814.1 | XP_017819303.1 | 2 - 3 | 997 | 98 | |