Assay Details
| Gene Symbol: RNF145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806589.4 | XP_002806635.2 | 5 - 6 | 700 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF145| Gene Symbol: RNF145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806589.4 | XP_002806635.2 | 5 - 6 | 700 | 71 | |
Set Membership: |
|