Assay Details
| Gene Symbol: RNPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755798.4 | XP_002755844.3 | 5 - 6 | 693 | 62 | |
| XM_017978632.1 | XP_017834121.1 | 6 - 7 | 881 | 62 | ||
| XM_017978633.1 | XP_017834122.1 | 6 - 7 | 1771 | 62 | ||
| XM_017978634.1 | XP_017834123.1 | 6 - 7 | 1007 | 62 | ||
| XM_017978635.1 | XP_017834124.1 | 6 - 7 | 915 | 62 | ||
More Information
Set Membership: |
|

RNPS1
Genomic Map