Assay Details
| Gene Symbol: GTF2F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749026.4 | XP_002749072.1 | 5 - 6 | 577 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GTF2F2| Gene Symbol: GTF2F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749026.4 | XP_002749072.1 | 5 - 6 | 577 | 70 | |
Set Membership: |
|