Assay Details
| Gene Symbol: CFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753811.3 | XP_002753857.1 | 3 - 4 | 659 | 87 | |
| XM_009005921.1 | XP_009004169.1 | 3 - 4 | 536 | 87 | ||
More Information
Set Membership: |
|

CFL2
Genomic Map