Assay Details
| Gene Symbol: PGBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993760.2 | XP_008992008.1 | 3 - 4 | 978 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PGBD1| Gene Symbol: PGBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993760.2 | XP_008992008.1 | 3 - 4 | 978 | 65 | |
Set Membership: |
|