Assay Details
| Gene Symbol: RAP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751223.3 | XP_002751269.1 | 7 - 8 | 808 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAP1A| Gene Symbol: RAP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751223.3 | XP_002751269.1 | 7 - 8 | 808 | 77 | |
Set Membership: |
|