Assay Details
| Gene Symbol: NAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745275.3 | XP_002745321.1 | 1 - 2 | 549 | 85 | |
| XM_008992522.2 | XP_008990770.1 | 1 - 2 | 549 | 85 | ||
More Information
Set Membership: |
|

NAF1
Genomic Map