Assay Details
| Gene Symbol: YDJC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978214.1 | XP_017833703.1 | 1 - 2 | 221 | 103 | |
| XM_017978215.1 | XP_017833704.1 | 1 - 2 | 236 | 103 | ||
| XM_017978219.1 | XP_017833708.1 | 1 - 2 | 475 | 103 | ||
More Information
Set Membership: |
|

YDJC
Genomic Map