Assay Details
| Gene Symbol: LOC100398456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987600.2 | XP_008985848.2 | 1 - 2 | 876 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008987600.2 transcripts.
Set Membership: |
|

LOC100398456
Genomic Map