Assay Details
| Gene Symbol: UBQLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742796.3 | XP_002742842.1 | 10 - 11 | 2177 | 76 | |
| XM_008994579.2 | XP_008992827.1 | 9 - 10 | 2098 | 76 | ||
| XM_017972584.1 | XP_017828073.1 | 10 - 11 | 1622 | 76 | ||
More Information
Set Membership: |
|

UBQLN1
Genomic Map