Assay Details
| Gene Symbol: AMACR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017969966.1 | XP_017825455.1 | 4 - 5 | 867 | 70 | |
| XM_017969967.1 | XP_017825456.1 | 3 - 4 | 675 | 70 | ||
More Information
Set Membership: |
|

AMACR
Genomic Map