Assay Details
| Gene Symbol: CAJA-DRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270911.1 | NP_001257840.1 | 1 - 2 | 180 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAJA-DRA| Gene Symbol: CAJA-DRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270911.1 | NP_001257840.1 | 1 - 2 | 180 | 100 | |
Set Membership: |
|