Assay Details
| Gene Symbol: RAB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742536.2 | XP_002742582.1 | 1 - 2 | 434 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB20| Gene Symbol: RAB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742536.2 | XP_002742582.1 | 1 - 2 | 434 | 59 | |
Set Membership: |
|