Assay Details
| Gene Symbol: ZFP69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974593.1 | XP_017830082.1 | 1 - 2 | 393 | 89 | |
| XM_017974594.1 | XP_017830083.1 | 1 - 2 | 393 | 89 | ||
More Information
Set Membership: |
|

ZFP69B
Genomic Map