Assay Details
| Gene Symbol: CRELD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743867.3 | XP_002743913.1 | 11 - 12 | 1313 | 68 | |
| XM_002743868.3 | XP_002743914.1 | 9 - 10 | 1172 | 68 | ||
More Information
Set Membership: |
|

CRELD2
Genomic Map