Assay Details
| Gene Symbol: AGRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761172.3 | XP_002761218.1 | 3 - 4 | 1235 | 116 | |
| XM_008986057.2 | XP_008984305.1 | 4 - 5 | 643 | 116 | ||
More Information
Set Membership: |
|

AGRP
Genomic Map