Assay Details
| Gene Symbol: TNNI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243071.1 | NP_001230000.1 | 6 - 7 | 406 | 66 | |
| XM_008985339.2 | XP_008983587.1 | 6 - 7 | 401 | 66 | ||
More Information
Set Membership: |
|

TNNI1
Genomic Map