Assay Details
| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753830.4 | XP_002753876.1 | 3 - 4 | 1717 | 77 | |
| XM_017977058.1 | XP_017832547.1 | 1 - 2 | 91 | 77 | ||
More Information
Set Membership: |
|

SLC25A21
Genomic Map