Assay Details
| Gene Symbol: NFKBIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987961.2 | XP_008986209.1 | 714 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NFKBIB| Gene Symbol: NFKBIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987961.2 | XP_008986209.1 | 714 | 94 | ||