Assay Details
| Gene Symbol: C1H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742790.4 | XP_002742836.1 | 1 - 2 | 576 | 72 | |
| XM_017972554.1 | XP_017828043.1 | 2 - 3 | 232 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017972554.1 transcripts.
Set Membership: |
|

C1H9orf64
Genomic Map