Assay Details
| Gene Symbol: C1H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742790.4 | XP_002742836.1 | 2 - 3 | 843 | 91 | |
| XM_017972554.1 | XP_017828043.1 | 3 - 4 | 499 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1H9orf64| Gene Symbol: C1H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742790.4 | XP_002742836.1 | 2 - 3 | 843 | 91 | |
| XM_017972554.1 | XP_017828043.1 | 3 - 4 | 499 | 91 | ||