Assay Details
| Gene Symbol: C1H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742790.4 | XP_002742836.1 | 3 - 4 | 1036 | 66 | |
| XM_017972554.1 | XP_017828043.1 | 4 - 5 | 692 | 66 | ||
More Information
Set Membership: |
|

C1H9orf64
Genomic Map